Key Documents
PCR96-AB-C-10
Novas Bio
Novas Bio 96-well PCR Plate
10 Plates, 0.2 ml 96-well PCR plate. Semi-skirted. Elevated skirt for ABI.
About This Item
PDF Icon
Shipped & Sold By Novas Bio
A Trusted Marketplace Seller
PDF Icon
Have a question?
material
polypropylene
size
96 wells
General description
  • 96 well x 0.2 ml. Standard profile
  • Single notch corner at A12
  • Especially designed for ABI fast cyclers and sequencers:
    • Raised wells for optimum seal and minimized evaporation
    • Elevated skirt around plate provides optimum fit with ABI blocks
    • Semi-skirted design allows for labeling
    • Manufactured with medical-grade polypropylene
    • Thin wall design for efficient heat transfer
    • Exceptional clarity for maximum sample visibility
    • Certified DNase, RNase, Human DNA free. Non-pyrogenic.
    • Pairs perfectly with Novas Bio sequencing mat
Choose from one of the most recent versions:
Certificates of Analysis (COA)