Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y04T-04-5PK

CELLASIC® ONIX Pad Trap Plate (4-chamber, trap heights 4 micron), 5pk

Synonym(s):

4-Chamber Trap Plate, ONIX Pad Trap Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161500

material

PMMA
glass
polycarbonate
polydimethylsiloxane (PDMS)

packaging

pack of 5 ea

L × W × H

6.0 mm × 2.3 mm × 4.0 μm

General description

Barrier structures retain cells while enabling perfusion to wash away smaller cells.
Cell size range: 3.5-5.0 μm
Device configuration: 4 chamber

Application

The Y04T pad trap plates for 4 μm cells utilize microfabricated PDMS pillars that extend from the ceiling to selectively trap cells and restrict their growth in a single focal plane & maintain x,y positioning over time.

Legal Information

CELLASIC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service