Journal of food science and technology, 57(1), 22-31 (2020-01-25)
The coconut whey (CW) (underutilized by-product of virgin coconut oil production, with 2-3% protein) was subjected to ultrafiltration for concentration of protein and sugar removal prior to spray drying. The process parameters of ultrafiltration were standardised with respect to transmembrane
Questions
Reviews
★★★★★ No rating value