Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX0143J

Supelco

Silica Gel Grade 12

28-200 Mesh, Meets ACS Specifications GR ACS

Synonym(s):

Silica gel, Amorphous Silicon Dioxide, Amorphous Silica

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 KG
$449.00

$449.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Select a Size

Change View
2.5 KG
$449.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41123003
EC Index Number:
231-545-4

$449.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

form

powder or crystals

color

white

particle size

<2 mm
28-200 mesh (>=80%)

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

shipped in

ambient

storage temp.

room temp

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Color: White
Form: Crystals to powder
Mesh size (28-200): 80% min
Suitability for moisture absorption: To pass test

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service