Skip to Content
MilliporeSigma
All Photos(2)

Documents

SCT251

Sigma-Aldrich

BioTracker BSH Activity Bioluminescent Probe

Human

Synonym(s):

Bile Salt Hydrolase probe;BSH probe;BSH bioluminescent probe;BSH cell and tissue probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.47

product name

BioTracker BSH Activity Bioluminescent Probe,

biological source

human

Quality Level

form

lyophilized

packaging

pkg of 1 mg

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

activity assay: suitable
cell based assay: suitable
enzyme immunoassay: suitable
microbiological culture: suitable

detection method

bioluminescent

shipped in

dry ice

storage temp.

−20°C

General description

Reference
Sci Adv. 2021, 7(6): eaaz9857.

Application

Purity: > 97% confirmed by HPLC, HNMR, LC-MS and elemental analysis. Molar Mass: 653.89 g/mol

Features and Benefits

This BSH-activatable luciferin (BAL) probe provides a noninvasive optical readout-based method for quantifying BSH activity. Biological applications include measurement of BSH activity of pure enzyme, or diverse samples such as live bacteria, fecal samples, or through the use of in vivo noninvasive imaging.

Physical form

Lyophilized. Yellow solid.

Storage and Stability

Store BioTracker Bile Salt Hydrolase Bioluminescent Probe at -20°C, desiccated and protected from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Other Notes

Bioluminescent enzyme activity measurement in cells and tissues

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service