Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R03131

Sigma-Aldrich

Safranin Solution

Harleco®, 1%, aqueous solution, suitable for histology

Synonym(s):

Gram Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

form

aqueous solution

Quality Level

manufacturer/tradename

Harleco®

concentration

1%

application(s)

diagnostic assay manufacturing
hematology

histology

shipped in

ambient

storage temp.

15-25°C

General description

HARLECO® 1% aqueous solution for general purpose counterstaining
Safranin Solution
R03131-74 500ml Poly Bottle

Application

HARLECO® 1% aqueous solution for general purpose counterstaining
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Aqueous solution for general purpose counterstaining.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service