MAPK8960C
MultiScreen Punch Kit for 12 x 75 mm tubes
Radionucleotide counting by transfer of individual filters to tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product line
MultiScreen®
manufacturer/tradename
MultiScreen®
shipped in
ambient
General description
MultiScreen Punch Kit and Accessories
Patented punch tips remove each filter from its well separately, thereby eliminating the risk of cross-contamination.
The MultiScreen Punch consists of a punch handle, a sliding punch carrier plate, a punch distributor, and an eight-place carrier rack to hold samples.
After completing your assay, you can use the punch and disposable punch tips to simultaneously remove filters from eight wells of a MultiScreen plate and distribute them into eight scintillation vials for scintillation or gamma radioisotope detection.
The patented punch tips remove each filter from its well separately, thereby eliminating the risk of cross-contamination and increasing reproducibility.
Patented punch tips remove each filter from its well separately, thereby eliminating the risk of cross-contamination.
The MultiScreen Punch consists of a punch handle, a sliding punch carrier plate, a punch distributor, and an eight-place carrier rack to hold samples.
After completing your assay, you can use the punch and disposable punch tips to simultaneously remove filters from eight wells of a MultiScreen plate and distribute them into eight scintillation vials for scintillation or gamma radioisotope detection.
The patented punch tips remove each filter from its well separately, thereby eliminating the risk of cross-contamination and increasing reproducibility.
Components
Multiple punch
1 carrier rack for 12 x 75 mm tubes
1 carrier rack for 12 x 75 mm tubes
Legal Information
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service