MABS1262
Anti-phospho-ZIP7 (Ser275/276) Antibody, clone 3-3-3
clone 3-3-3, from mouse
Synonym(s):
Zinc transporter SLC39A7, S256/S257-phosphorylated, Histidine-rich membrane protein Ke4, S256/S257-phosphorylated, Really interesting new gene 5 protein, S256/S257-phosphorylated, Solute carrier family 39 member 7, S256/S257-phosphorylated, ZIP7, S256/S2
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
3-3-3, monoclonal
species reactivity
human
species reactivity (predicted by homology)
nonhuman primates (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology), sheep (based on 100% sequence homology), canine (based on 100% sequence homology)
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
phosphorylation (pSer275/pSer276)
Gene Information
human ... SLC39A7(7922)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: Clone 3-3-3 hybridoma culture supernatant (1:1,000 dilution) detected a ~48 kDa phosphorylated target band in lysate from wild-type ZIP7-transfected MCF-7 cells (Courtesy of Kathryn Taylor, Ph.D., Cardiff University, United Kingdom).
Signaling
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected ZIP7 S256/S257 phosphorylation in 20 µg of MCF-7 cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service