MABN694
Anti-CHD3 Antibody, clone 2G4
ascites fluid, clone 2G4, from mouse
Synonym(s):
Chromodomain-helicase-DNA-binding protein 3, ATP-dependent helicase CHD3, Mi-2 autoantigen 240 kDa protein, Mi2-alpha, Zinc finger helicase, hZFH, CHD3
About This Item
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
2G4, monoclonal
species reactivity
mouse, human
technique(s)
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CHD3(1107)
General description
Immunogen
Application
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected CHD3 in HeLa cells.
Flow cytometry analysis: A 1:200-400 dilution from a representative lot detected CHD3 in K562 cells.
Optimal working dilutions must be determined by end user.
Neuroscience
Developmental Neuroscience
Quality
Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected CHD3 in mouse brain, HeLa, K562, Jurkat, NTERA-2, HEK293, Raji lysates.
Target description
Linkage
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
mouse brain, HeLa, K562, Jurkat, NTERA-2, HEK293, Raji lysates
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service