Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN373

Sigma-Aldrich

Anti-Neuroleukin/GPI Antibody, clone 10C2.1

clone 10C2.1, from mouse

Synonym(s):

Zinc finger FYVE domain-containing protein 26, FYVE domain-containing centrosomal protein, FYVE-CENT, Spastizin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10C2.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZFYVE26(23503)

General description

Glucose-6-phosphate isomerase (GPI) alternatively known as Autocrine motility factor (AMF), Neuroleukin (NLK), Phosphoglucose isomerase (PGI), Phosphohexose isomerase (PHI) or Sperm antigen 36 (SA-36) and encoded by the gene name GPI is major enzyme in glycolysis and gluconeogenesis. Glucose-6-phosphate isomerase rearranges glucose-6-phosphate (from the phosphorylation of glucose via hexokinases) into fructose-6-phosphate (F6P) as the second step in glycolysis. Interestingly however, Glucose-6-phosphate isomerase plays an entirely different sort of role outside of the cell than when it is inside being part of glycolysis. When secreted, Glucose-6-phosphate isomerase acts as an autocrine motility factor for various cancers and is critical for metastasis, while when secreted in nervous tissue it acts as a neurotrophic factor for sensory and spinal neurons and in lymphocytes it acts as a lymphokine upon T cell ligand binding and stimulates B cells to secrete immunoglobulin. Mutations in Glucose-6-phosphate isomerase are the second most frequent cause of inherited glycolytic-enzymopathy in humans. This autosomal recessive disorder is characterized by a non-spherocytic anemia of variable severity which can present with neuromuscular dysfunctions defined by muscle weakness and mental retardation.

Immunogen

GST-tagged recombinant protein corresponding to human Neuroleukin/GPI.

Application

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Neuroleukin/GPI in human thalamus and rat hindbrain tissues.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience
This Anti-Neuroleukin/GPI Antibody, clone 10C2.1 is validated for use in western blotting & IHC (Paraffin) for the detection of Neuroleukin/GPI.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 μg/mL of this antibody detected Neuroleukin/GPI in 10 μg of HeLa cell lysate.

Target description

~58 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service