Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN1182

Sigma-Aldrich

Anti-Glutamine Synthetase Antibody, clone 11A3.1

clone 11A3.1, from mouse

Synonym(s):

Glutamine synthetase, Glutamate-ammonia ligase, Glutamate decarboxylase, GS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11A3.1, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GLUL (2752)
rat ... Glul(24957)

General description

Glutamine synthetase (EC 4.1.1.15, EC 6.3.1.2; UniProt P09606; also known as Glutamate-ammonia ligase, Glutamate decarboxylase, GS) is encoded by the Glul (also known as Glns) gene (Gene ID 24957) in rat species. Glutamine synthetase (GS) plays an essential role in nitrogen metabolism by catalyzing ATP-dependent condensation of glutamate and ammonia to form glutamine. Three classes of GS exist, class I enzymes (GSI) are specific to prokaryotes and exist as oligomers of 12 identical subunits, class II enzymes (GSII) exist as decamer of identical subunits in eukaryotes and in some bacteria, while class III enzymes (GSIII) function as a double-ringed dodecamer of identical chains found in bacteroides fragilis and in butyrivibrio fibrisolvens. GS is present predominantly in the brain, kidneys, and liver in mammals. In the brain, GS is found primarily in astrocytes, where it participates in the the detoxification of ammonia via assimilation and termination of neurotransmitter signals via recyclization. c-Myc promotes glutamine usage by upregulating glutaminase (GLS), which converts glutamine to glutamate in the TCA cycle, while in some cancer cells Myc-mediated thymine DNA glycosylase (TDG) transcription leads to GS upregulation due to TDG-dependent demethylation of the GS promoter. GS upregulation promotes cancer cell survival under glutamine limitation, while GS silencing decreases cancer cell proliferation and xenograft tumor growth.

Specificity

Clone 11A3.1 targets an internal epitope within the C-terminal half of human and rat glutamine synthetase.

Immunogen

GST-tagged recombinant rat glutamine synthetase internal fragment.

Application

Anti-Glutamine Synthetase, clone 11A3.1, Cat. No. MABN1182, is a highly specific mouse monoclonal antibody that targets Glutamine Synthetase, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A 1:20,000 dilution of from a representative lot detected Glutamine synthetase in 10 µg of human brain tissue lysate.

Immunohistochemistry Analysis: A 1:1000 dilution from a representative lot detected Glutamine synthetase in human cerebellum, cerebral cortex, and tonsil tissue sections.

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: A 1:20,000 dilution of this antibody detected Glutamine synthetase in 10 µg of rat brain tissue lysate.

Target description

~42 kDa observed. 42.06 kDa (human) and 42.27 kDa (rat) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid Zayas-Santiago et al.
International journal of molecular sciences, 25(12) (2024-06-27)
Polyamine (PA) spermidine (SPD) plays a crucial role in aging. Since SPD accumulates in glial cells, particularly in Müller retinal cells (MCs), the expression of the SPD-synthesizing enzyme spermidine synthase (SpdS) in Müller glia and age-dependent SpdS activity are not
Houjian Zhang et al.
NPJ Regenerative medicine, 8(1), 36-36 (2023-07-14)
Mammalian Müller glia (MG) possess limited regenerative capacities. However, the intrinsic capacity of mammalian MG to transdifferentiate to generate mature neurons without transgenic manipulations remains speculative. Here we show that MAP4K4, MAP4K6 and MAP4K7, which are conserved Misshapen subfamily of
David S Ríos et al.
Biomedicines, 11(4) (2023-05-16)
Polyamines (PAs) in the nervous system has a key role in regeneration and aging. Therefore, we investigated age-related changes in the expression of PA spermidine (SPD) in the rat retina. Fluorescent immunocytochemistry was used to evaluate the accumulation of SPD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service