Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE364

Sigma-Aldrich

Anti-H2A (H4R3me2s) Antibody, clone 6G1/1D3

clone 6G1/1D3, from mouse

Synonym(s):

Histone H2A type 1, H2A.1, Histone H2A/p+C10, Anti-H2A (H4R3me2s)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G1/1D3, monoclonal

species reactivity

human

technique(s)

dot blot: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

dimethylation (Arg3)

Gene Information

General description

Histone H2A type 1 (UniProt: P0C0S8; also known as H2A.1, Histone H2A/p) is encoded by the HIST1H2AG (also known as HIST1H2AG, H2AFP, HIST1H2AI, H2AFC) gene (Gene ID: 8329, 8320, 8332, 8336, 8369) in human. Histone H2A is one of four components of the core nucleosomal structure. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Histones undergo a number of post-translational modifications (PTM) in response to various stimuli that serve important functions for regulation of gene expression, DNA repair, and chromatin. Histone H2A, in particular, undergoes acetylation on selected lysines, which may promote unfolding of chromatin and transcription. It can also be phosphorylated on selected serine residues, which mark mitosis and gene repression, or DNA damage. Arginine methylation is a less explored histone post-translational modification. It has been implicated in embryonic development, somatic transcriptional regulation, and cancer development. H2A arginine 3 methylation (R3me1, R3me2s, and R3me2a) is catalyzed by PRMT1 and PRMT5 in vitro. H2A methylation is shown to appear late in oogenesis and is most abundant in the laid egg. Arginine methylations are constrained to pre-mid blastula transition events in the embryo and are considered to be involved in the global transcriptional repression during development. H2A R3me2s is reported to be enriched on nucleosomes containing both active and repressive histone post-translational modifications in human A549 cells and Xenopus embryos. (Ref.: Wilczek, C et al. (2011). J Biol Chem. 286(49):42221-31; Wang, WL et al. (2014). BMC Epigenetics & Chromatin 7:22).

Specificity

Clone 6G1/1D3 specifically detects H2A/H4 arginine 3 symmetric dimethylation (H2A/H4R3me2) in acid extract of HeLa cells.


Immunogen

KLH-conjugated linear symmetric di methyl R3 peptide.

Application

Anti-H2A (H4R3me2s), clone 6G1/1D3, Cat. No. MABE364, is a highly specific mouse monoclonal antibody that targets H2A/H4R3me2s and has been tested in Dot Blot and Western Blotting.
Dot Blot Analysis: 1:500 dilution from a representative lot detected H2A (H4R3me2s) in H2A, H2B, and H4 dot blot.


Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cell acid extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected H2A (H4R3me2s) in 10 µg of HeLa cell acid extract.

Target description

~15 kDa observed; 14.09 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service