Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE1918

Sigma-Aldrich

Anti-TAF-15 Antibody, clone 12F11

clone 12F11, from rat

Synonym(s):

TATA-binding protein-associated factor 2N, 68 kDa TATA-binding protein-associated factor, TAF(II)68, TAFII68, RNA-binding protein 56

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12F11, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2cκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... TAF15(8148)

General description

TATA-binding protein-associated factor 2N (UniProt: Q92804; also known as 68 kDa TATA-binding protein-associated factor, TAF(II)68, TAFII68, RNA-binding protein 56, TAF15) is encoded by the TAF15 (also known as RBP56, TAF2N) gene (Gene ID: 8148) in human. TAF15 is a ubiquitously distributed protein that is found in all fetal and adult tissues. It is a RNA and ssDNA-binding protein that plays specific roles during transcription initiation at distinct promoters and can enter the preinitiation complex together with the RNA polymerase II (Pol II). TAF15 shuttles between the nucleus and the cytoplasm. It can be dimethylated by protein arginine N-methyltransferase 1 (PRMT1) at Arg206 to asymmetric dimethylarginine. This methylation favors its nuclear localization and it also considered to be essential for its gene regulatory function. TAF15 contains an N-terminal activation domain, an RNA recognition motif (RRM) and many Arg-Gly-Gly (RGG) repeats at its C-terminal end. Its N-terminus serves as an essential transforming domain in the fusion oncoprotein created by chromosomal translocation in certain human chondrosarcomas. Depletion of TAF15 has a growth-inhibitory effect and results in increased apoptosis. (Ref.: Ballarino, M et al. (2013). Oncogene 32(39); 4646-4655; Jobert, L., et al. (2008). Exp. Cell res. 315(7);1273-1286).

Specificity

Clone 12F11 is a rat monoclonal antibody that detects human TATA-binding protein-associated factor 2N (TAF15). It targets an epitope with in 20 amino acids from the C-terminal half.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 20 amino acids from the C-terminal half of human TATA-binding protein-associated factor 2N (TAF15).

Application

Anti-TAF-15, clone 12F11, Cat. No. MABE1918, is a highly specific rat monoclonal antibody that targets TATA-binding protein-associated factor 2N and has been tested for use in ELISA and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected TAF-15 in Western Blotting applications (Suarez-Calvet, M., et. al. (2016). Acta Neuropathol. 131(4):587-604).

ELISA Analysis: A representative lot detected TAF-15 in ELISA applications (Suarez-Calvet, M., et. al. (2016). Acta Neuropathol. 131(4):587-604).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected TAF-15 in HEK293 cell lysate.

Target description

~68 kDa observed; 61.83 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2c in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service