Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MABE1068

Sigma-Aldrich

Anti-APE/Ref-1 Antibody, clone 13B8E5C2

clone 13B8E5C2, from mouse

Synonym(s):

DNA-(apurinic or apyrimidinic site) lyase, AP endonuclease 1, APE-1, APEN, APEX nuclease, Apurinic-apyrimidinic endonuclease 1, Redox factor-1, REF-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$414.00

$414.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$414.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$414.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13B8E5C2, monoclonal

species reactivity

human, rat, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HAP1(9001)

General description

DNA-(apurinic or apyrimidinic site) lyase (EC 4.2.99.18; UniProt P27695; also known as AP endonuclease 1, APE-1, APEN, APEX nuclease, Apurinic-apyrimidinic endonuclease 1, Redox factor-1, REF-1) is encoded by the APEX1 (also known as APE, APE1, APEX, APX, HAP1, REF1) gene (Gene ID 328) in human. APE/Ref-1 mediates the cleavage of an apurinic/apyrimidinic (AP) site in DNA generated by DNA glycosylases during base excision repair (BER) in both nucleus and mitochondria. APE/Ref-1 also functions as a redox factor (ref) responsible for maintaining several transcription factors (TFs) in an active reduced state for DNA binding, including AP-1, NF-κB, HIF-1α, CREB, TP53. Elevated levels of APE1 are associated with resistance to chemotherapyand poor prognosis in various cancers. APE/Ref-1 is synthesized as a 318-a.a. protein whose initiation methionine (Met1) is removed posttranslationally to yield the 317-a.a. form with a nuclear localization sequence (NLS; a.a. 8-13) and a nuclear export signal (NES; a.a. 64-80) sequence to allow its shuttle between the cytoplasm and nucleus. The 317-a.a. form can be further processed by a mitochondria-associated N-terminal peptidase into a mitochrondrial form (mtAPE; a.a. 32-318) that lacks NLS and contains only the NES and the mitochondrial targeting sequence (MTS; a.a. 289-318).

Specificity

Clone 13B8E5C2 detected both cytosolic and nuclear APE/Ref-1 by Immunocytochemistry, Immunohistochemistry, and Western blotting (Moore, D.H., et al. (2000). Clin. Cancer Res. 6(2):602-609).

Immunogen

Recombinant full-length human APE/Ref-1.

Application

Anti-APE/Ref-1, clone 13B8E5C2, Cat. No. MABE1068, is a highly specific mouse monoclonal antibody, that targets APE1 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:200 dilution from a representative lot detected both cytosolic and nuclear APE/Ref-1 immunoreactivity in 4% paraformaldehyde-fixed, 0.3% Triton X-100-permeabilized HeLa cells.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected nuclear APE/Ref-1 immunoreactivity in rat kidney, and human placenta & small intestine tissue sections.

Western Blotting Analysis: 0.5 µg/mL from a representative lot detected APE/Ref-1 in 10 µg of Raji cell lysate.

Immunocytochemistry Analysis: A representative lot detected both cytosolic and nuclear APE/Ref-1 immunoreactivity in HeyC2 and HEY human ovarian cancer cell lines (Moore, D.H., et al. (2000). Clin. Cancer Res. 6(2):602-609).

Immunohistochemistry Analysis: A representative lot detected varying tissue distribution and cellular localization of APE/Ref-1 immunoreactivity among various 4% formaldehyde-fixed, paraffin-embedded human ovarian tissue samples, including ovarian endometriosis, serous cystadenoma , serous carcinoma, ands well as normal ovarian tissues (Moore, D.H., et al. (2000). Clin. Cancer Res. 6(2):602-609).

Western Blotting Analysis: A representative lot detected an upregulated cytosolic APE/Ref-1 level in HeyC2 than HEY cells, while similar nuclear APE/Ref-1 level was detected in these two human ovarian cancer cell lines (Moore, D.H., et al. (2000). Clin. Cancer Res. 6(2):602-609).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected APE/Ref-1 in 10 µg of HepG2 cell lysate.

Target description

~35-39 kDa observed. 35.42 kDa (Met1 removed; a.a. 2-318) and 32.24 kDa (Mitochondrial form; a.a. 32-318) calculated. The broad banding pattern is consistent with the detection of both mitochondrial and non-mitochondrial forms with varying degrees of posttranslational modifications (lysine acetylation, threonine phosphorylation, cysteine nitrosylation). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service