Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABD131

Sigma-Aldrich

Anti-ISL1 Antibody, clone 1H9

ascites fluid, clone 1H9, from mouse

Synonym(s):

Insulin gene enhancer protein ISL-1, Islet-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1H9, monoclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ISL1(3670)

General description

Islet-1 (ISL-1) is a transcription factor that plays an important role in the embryogenesis of pancreatic islets of Langerhans. It is expressed in subsets of neurons of the adrenal medulla and dorsal root ganglion, inner nuclear and ganglion cell layers in the retina, the pineal and some regions of the brain. In mouse embryos, a deficiency of this gene fail to undergo neural tube motor neuron differentiation. More recently it has been identified as a marker for a cardiac progenitor cell lineage that is capable of differentiating into all 3 major heart cell types: cardiomyocytes, smooth muscle and endothelial cell lineages.

Immunogen

Purified recombinant fragment of human ISL1 expressed in E. Coli. corresponding to human ISL1.

Application

Detect ISL1 using this mouse monoclonal antibody, Anti-ISL1 Antibody, clone 1H9 validated for use in western blotting, Immunofluorescence & IHC.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected ISL1 in full-length ISL1-hIgGFc transfected HEK293 cells.

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected ISL1 in paraffin-embedded human lung cancer and cervical carcinoma tissues.

Optimal working dilutions must be determined by end user.

Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in full-length ISL1-hIgGFc transfected HEK293 cell lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected ISL1 in full-length ISL1-hIgGFc transfected HEK293 cell lysate.

Target description

~93 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Full-length ISL1-hIgGFc transfected HEK293 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service