Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB2150

Sigma-Aldrich

Anti-E-Selectin Antibody, clone P2H3

clone P2H3, Chemicon®, from mouse

Synonym(s):

ELAM-1, CD62E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

P2H3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SELE(6401)

General description

Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface, members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNF alpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NK kappa B dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.

Specificity

Human E-Selectin

Immunogen

IL-1(1 activated HUVEC (human)

Application

Anti-E-Selectin Antibody, clone P2H3 detects level of E-Selectin & has been published & validated for use in ELISA, FC, IP, IC, IH.
Immunohistochemistry: 1:50 – 1:500; acetone or paraformaldehyde-fixed frozen and formalin-fixed paraffin-embedded tissues; recommend heat-induced epitope retrieval in EDTA, pH 8.0 for paraffin-embedded tissues
Immunofluorescence
Flow Cytometry
Immunoprecipitation
ELISA
Blocking: 10-15µg/mL; inhibits carcinoma and monocyte adhesion to IL-1β activated HUVEC

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified by protein G and stored in 0.1 M Tris-Glycine, pH 7.4, 150 mM NaCl containing 0.05% NaN3.

Storage and Stability

Maintain at 2-8°C for up to 12 months from date of receipt..

Analysis Note

Control
Ductal breast cancer or mormal colon

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalya V Narizhneva et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 19(9), 1158-1160 (2005-04-19)
Expression of cell adhesion molecules (CAM) responsible for leukocyte-endothelium interactions plays a crucial role in inflammation and atherogenesis. Up-regulation of vascular CAM-1 (VCAM-1), intracellular CAM-1 (ICAM-1), and E-selectin expression promotes monocyte recruitment to sites of injury and is considered to
Zheng-Gen Jin et al.
Arteriosclerosis, thrombosis, and vascular biology, 24(7), 1186-1191 (2004-05-08)
Cyclophilin A (CyPA) is an abundant intracellular protein that is considered to be the main target of the immunosuppressive drug cyclosporine A. We and others showed that CyPA is secreted from smooth muscle cells and macrophages in response to oxidative
Hila Israelov et al.
Journal of neuroinflammation, 17(1), 267-267 (2020-09-11)
Excessive inflammation might activate and injure the blood-brain barrier (BBB), a common feature of many central nervous system (CNS) disorders. We previously developed an in vitro BBB injury model in which the organophosphate paraoxon (PX) affects the BBB endothelium by
Cheikh I Seye et al.
The Journal of biological chemistry, 278(27), 24960-24965 (2003-04-26)
P2Y2 receptor up-regulation and activation induces intimal hyperplasia and monocyte/macrophage infiltration in the collared rabbit carotid artery model of vascular injury, suggesting a potential role for P2Y2 receptors in monocyte recruitment by vascular endothelium. In this study, we addressed the
Gahee Min et al.
Journal of cellular and molecular medicine, 20(12), 2333-2340 (2016-07-16)
Drug repositioning is used to discover drug candidates to treat human diseases, through the application of drugs or compounds that are approved for the treatment of other diseases. This method can significantly reduce the time required and cost of discovering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service