Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS207RTA

Sigma-Aldrich

Ready-to-Assay LPA2 Lysophospholipid Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.84
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

Product Name

Ready-to-Assay LPA2 Lysophospholipid Receptor Frozen Cells,

biological source

human

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Lysophosphatidic acid (LPA) is a lysophospholipid produced by activated platelets that inhibits adenylate cyclase and stimulates DNA synthesis, changes in cell morphology, and increases in intracellular calcium in a variety of cultured mammalian cells. A family of three GPCRs, LPA1, LPA2, and LPA3, mediates the biological effects of LPA (Contos et al., 2000). LPA2 is widely expressed, with particularly notable expression in helper T Cells (Huang et al.) LPA2 upregulation has been demonstrated in multiple cancer cells, including colon and ovarian cancer (Yun et al.). LPA2 couples to multiple signaling cascades through Gq Gi/o and G12/13, mediating inhibition of adenyl cyclase, inositol phosphate turnover, calcium flux and cell migration Millipore′s cloned human LPA2 expressing cell line is made in the Chem-1 host, which supports high levels of recombinant LPA2 expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at LPA2.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human LPA2 Lysophospholipid GPCR Frozen Cells for calcium flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: LPA2
  • Target Sub-Family: Lysophospholipid

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service