HTS138M
ChemiSCREEN Membrane Preparation Recombinant Human CXCR7 Chemokine Receptor
Human CXCR7 / RDC1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
biological source
human
Quality Level
recombinant
expressed in Chem-1 cells
manufacturer/tradename
ChemiScreen
Chemicon®
technique(s)
ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
General description
: Full-length human CXCR7 cDNA encoding CXCR7
The orphan receptor RDC1 is phylogenetically related to chemokine receptors, and its gene (CMKOR1, chemokine orphan receptor 1) occupies a chromosomal location near the genes for other CXC receptors. Several immune system cells, including T lymphocytes, mature dendritic cells and memory B cells, express RDC1 (Infantino et al., 2006). A subset of HIV and SIV strains use RDC1 as a coreceptor (Shimizu et al., 2000). The chemokine SDF-1/CXCL12, which binds to CXCR4, has been shown also to bind and activate RDC1 to promote chemotaxis of T lymphocytes; thus, CXCR7 has been proposed to be the new name for RDC1 (Balabanian et al., 2005). I-TAC, one of the chemokines thought to bind to CXCR3 only, has been reported to distinguish CXCR7 from CXCR4 in radioligand binding experiments (Burns et al. 2006). Chemicon′s CXCR7 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of agonists and antagonists at CXCR7. The membrane preparations exhibit a Kd of 0.32 nM for [125I]-SDF-1α. With 0.25 nM [125I]-SDF-1α, 10µg/well CXCR7 Membrane Prep typically yields greater than 4-fold signal-to-background ratio.
Application
Radioligand binding assay and GTPγS binding
Biochem/physiol Actions
GPCR Class: A
Protein Target: CXCR7 / RDC1
Target Sub-Family: Chemokine
Quality
Table 1. Signal:background and specific binding values obtained in a competition binding assay with CXCR7 membrane prep and unlabeled SDF-1α.
1 unit = 10 µg
Bmax for [125I]- SDF-1α binding: 0.51 pmol/mg protein
Kd for [125I]- SDF-1α binding: ~0.32 nM
10 µg/well | |
---|---|
Signal:Background | 4.92 |
Specific Binding (cpm) | 15919 |
1 unit = 10 µg
Bmax for [125I]- SDF-1α binding: 0.51 pmol/mg protein
Kd for [125I]- SDF-1α binding: ~0.32 nM
Specifications
Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I] SDF-1α (Perkin Elmer#: NEX-346)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 4-fold signal:background with 125I-labeled SDF-1α at 0.25 nM
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I] SDF-1α (Perkin Elmer#: NEX-346)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 4-fold signal:background with 125I-labeled SDF-1α at 0.25 nM
Physical form
Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membranes protein were adjusted to 1 mg/mL in packaging buffer, and dispensed at 1 mL/vial. Vials were rapidly frozen, and stored at -80°C.
Packaging method: Membranes protein were adjusted to 1 mg/mL in packaging buffer, and dispensed at 1 mL/vial. Vials were rapidly frozen, and stored at -80°C.
Storage and Stability
Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Integrative biology : quantitative biosciences from nano to macro, 2(11-12), 680-686 (2010-09-28)
Chemokine CXCL12 promotes CXCR4-dependent chemotaxis of cancer cells to characteristic organs and tissues, leading to metastatic disease. This study was designed to investigate how cells expressing CXCR7 regulate chemotaxis of a separate population of CXCR4 cells under physiologic conditions in
The Journal of experimental medicine, 203(9), 2201-2213 (2006-08-31)
The chemokine stromal cell-derived factor (SDF-1; also known as chemokine ligand 12 [CXCL12]) regulates many essential biological processes, including cardiac and neuronal development, stem cell motility, neovascularization, angiogenesis, apoptosis, and tumorigenesis. It is generally believed that SDF-1 mediates these many
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service