Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HTS066M

Sigma-Aldrich

ChemiSCREEN Human Y 2 Neuropeptide Receptor Membrane Preparation

Human Y2 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
$884.00

$884.00


Estimated to ship onApril 08, 2025



Select a Size

Change View
200 UNITS
$884.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

$884.00


Estimated to ship onApril 08, 2025


biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... NPY2R(4887)

General description

Full-length human NPY2R cDNA encoding Y2
The NPY family consists of three 36-amino acid peptides, neuropeptide Y (NPY), peptide YY (PYY) and pancreatic polypeptide (PP), which bind to the NPY receptor family of G protein-coupled receptors. Five NPY receptors, Y1, Y2, Y4, Y5 and Y6, have been defined at the molecular level, and each signals primarily through Gi/o. Binding of NPY family peptides to NPY receptors mediates a variety of physiological effects, including promotion of food intake, decreased anxiety, inhibition of neurotransmitter and hormone release, vasoconstriction, and gut motility. Y2 is primarily expressed in the CNS, and it mediates presynaptic inhibition of neurotransmitter release (Michel et al., 1998). Chemicon′s Y2 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of Y2 interactions and its ligands. The membrane preparations exhibit a Kd of 1.28 nM for [125I]-human PYY. With 0.5 nM [125I]-human PYY, 5 or 10µg/well Y2 Membrane Prep yields greater than 10-fold signal-to-background ratio.

Application

Human Y2 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Radioligand binding assay and GTPγS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: Y2
Target Sub-Family: Neuropeptide Y

Quality

Signal:background and specific binding values obtained in a competition binding assay with varying amounts of TP membrane prep:
10 µg/well5 µg/well
Signal:Background 19.8 26.3
Specific Binding (cpm) 27347 23335


SPECIFICATIONS: 1 unit = 5 µg
Bmax for [125I] human PYY binding: 17.89 pmol/mg protein;
Kd for [125I] human PYY binding: ~ 1.28 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I]-human PYY (Perkin Elmer#:NEX-341 )
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 10-fold signal:background with 125I labeled human PYY at 0.5 nM

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.
Packaging method: Membranes protein were adjusted to 0.5 mg/ml in 1 mL packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

XVI. International Union of Pharmacology recommendations for the nomenclature of neuropeptide Y, peptide YY, and pancreatic polypeptide receptors.
M C Michel et al.
Pharmacological reviews, 50(1), 143-150 (1998-04-29)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service