Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB186F

Sigma-Aldrich

Milli-Mark Anti-CD20 -FITC Antibody, clone B-Ly1

clone B-Ly1, Milli-Mark®, from mouse

Synonym(s):

Membrane-spanning 4-domains subfamily A member 1

B-lymphocyte surface antigen B1

Leukocyte surface antigen Leu-16

Bp35

CD_antigen=CD20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

B-Ly1, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MS4A1(931)

General description

CD20 is a transmembrane, non-glycosylated protein expressed on B-cell precursors and mature B cells, but is lost following differentiation into plasma cells (Tedder, 1994). In resting B cells, CD20 appears in a 33 kDa non-phosphorylated form. After mitogen stimulation, CD20 becomes heavily phosphorylated (35-37 kDa isoforms), and it is a dominant phosphoprotein in activated B cells and B-cell lines (Zhou, 1995). The long N- and C-terminal ends of the protein are located on the cytoplasmic side of the membrane and only a minor portion of the protein is exposed on the cell surface (Tedder, 1994). Antibodies reacting with CD20 cytoplasmic epitopes are designated CD20cy (Zhou, 1995). It is suggested that CD20 plays a direct role in regulating the transmembrane conductive Ca 2+ flux of B cells which indicates a possible function for CD20 as a regulator of proliferation and differentiation (Tedder, 1994).

Specificity

Antibody recognizes Human CD20 antigen.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Milli-Mark Anti-CD20 -FITC Antibody, clone B-Ly1 is validated for use in FC for the detection of CD20.

Quality

Evaluated by flow cytometry using human lymphocytes.

Target description

33 kDa (Zhou, 1995)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1k conjugated to PE in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Human Lymphocytes

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service