B650-150S
Fluid Thioglycollate Medium (FTM)
with 0.5% Beef extract, bottle capacity 300 mL, suitable for sterility testing, bottle filling volume 150 mL, closure type, White screw cap with septum and protector, pack of 12 bottles
Synonym(s):
Fluid Thioglycollate Medium (FTM), FTM, Thioglycollate Broth
About This Item
Recommended Products
agency
EP 2.6.13
JP 4.06
USP 71
Quality Level
sterility
sterile; autoclaved
form
liquid
feature
closure type White screw cap with septum and protector
ready-to-use
composition
with 0.5% Beef extract
packaging
pack of 12 bottles
technique(s)
microbiological culture: suitable
sterility testing: suitable
bottle capacity
300 mL
bottle filling volume
150 mL
pH
6.9±0.2 (25 °C)
application(s)
food and beverages
pharmaceutical
storage temp.
2-25°C
suitability
nonselective for
Looking for similar products? Visit Product Comparison Guide
General description
The anaerobic atmosphere is created by reducing the redox potential of the medium by Thioglycollate and L-Cystine. Additionally, these compounds inactivate mercury and other heavy metal elements present in the environment. The presence of beef extract adds to the carbon source and other essential vitamins in the medium to support the growth of microorganisms present in the sample.
The presence of agar may reduce the rapid diffusion of oxygen and lead to slight turbidity in larger volumes (filled bottles). Resazurin indicator in the medium indicates the reduction potential. An increase in oxygen concentration is indicated through a color change reaction to pink.
Application
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service