Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AP160C

Sigma-Aldrich

Rabbit Anti-Mouse IgG Antibody, Cy3 conjugate

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

1.5 MG
$199.00

$199.00


Estimated to ship onMarch 20, 2025


Request a Bulk Order

Select a Size

Change View
1.5 MG
$199.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$199.00


Estimated to ship onMarch 20, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

CY3 conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reacts with Mouse IgG gamma chain, as well as the light chains from all Mouse immunoglobulin classes.

Monospecific on IEP against whole Mouse Serum.

FLOUROPHORE/PROTEIN RATIO:

A552/A280 = 2.46

Application

Detect Mouse IgG using this Rabbit anti-Mouse IgG Antibody, Cy3 conjugate validated for use in IF.

Storage and Stability

Store at +2-8°C. Do not freeze. After opening, restore to 1.1mL with distilled water. As an undiluted liquid the product is stable at 2-8°C for several weeks. For extended storage after reconstitution, we suggest the addition of an equal volume of glycerol to make a final glycerol concentration of 50% followed by storage at -20°C, with or without aliquoting. Exp. Date: 1 year from reconstitution.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurélie Allais-Bonnet et al.
FEBS open bio, 6(1), 4-15 (2016-04-06)
Three genes of the prion protein gene family are expressed in gonads. Comparative analyses of their expression patterns in mice and goats revealed constant expression of PRNP and SPRN in both species and in both male and female gonads, but
Golan Karvat et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(25) (2021-06-23)
The interaction between spontaneous and externally evoked neuronal activity is fundamental for a functional brain. Increasing evidence suggests that bursts of high-power oscillations in the 15- to 30-Hz beta-band represent activation of internally generated events and mask perception of external
Chizue Hiruta et al.
Zoological science, 29(11), 733-737 (2012-10-31)
In most animal species, centrosomes, the main microtubule-organizing centers, usually disintegrate in oocytes during meiosis and are reconstructed from sperm-provided centrioles before the first cleavage division. In parthenogenetic oocytes, however, no sperm-derived centrosome-dependent microtubule nucleation is expected, as fertilization does
Midori Awazu et al.
PloS one, 15(4), e0230289-e0230289 (2020-04-07)
We previously reported that maternal nutrient restriction (NR) inhibited ureteric branching, metanephric growth, and nephrogenesis in the rat. Here we examined whether folic acid, a methyl-group donor, rescues the inhibition of kidney development induced by NR and whether DNA methylation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service