Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN407

Sigma-Aldrich

Anti-GLUT9 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Solute carrier family 2, facilitated glucose transporter member 9, Glucose transporter type 9, GLUT-9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$377.00

$377.00


In Stock



Select a Size

Change View
100 μG
$377.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$377.00


In Stock


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC2A9(56606)

General description

GLUT9, also known as Glucose transporter type 9 or GLUT-9, and encoded by the gene SLC2A9/GLUT9, is a member of the sugar transport family that primarily transports urate and fructose; GLUT9 does also transport glucose but at a lower rate than its preferred sugars. GLUT9 also plays a role in urate reabsorption by the proximal tubules of the kidney. GLUT9 is localized to the plasma membrane. GLUT9 is most strongly expressed in basolateral membranes of proximal renal tubular cells, liver and placenta. GLUT9 is also detected in lung, blood leukocytes, heart skeletal muscle and chondrocytes from articular cartilage. Isoform 2 is only detected in the apical membranes of polarized renal tubular cells and placenta. Defects in GLUT9 may be associated with Hypouricemia renal 2 (RHUC2) a disorder characterized by impaired uric acid reabsorption at the apical membrane of proximal renal tubule cells, and high urinary urate excretion.

Specificity

This antibody is expected to recognize both known isoforms of GLUT9.

Immunogen

KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human GLUT9.

Application

Detect GLUT9 using this Anti-GLUT9 Antibody validated for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected GLUT9 in human cerebellum and hindbrain tissues.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected GLUT9 in 10 µg of human liver tissue lysate.

Target description

~62 kDa observed. Uniprot describes 2 isoforms at 59 kDa (isoform 1) and 55 kDa (isoform 2).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng Zhou et al.
Scientific reports, 14(1), 15449-15449 (2024-07-05)
Hyperuricemia (HUA), a metabolic disease caused by excessive production or decreased excretion of uric acid (UA), has been reported to be closely associated with a variety of UA transporters. Clerodendranthus spicatus (C. spicatus) is an herbal widely used in China

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service