Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN342

Sigma-Aldrich

Anti-FEZ2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Fasciculation and elongation protein zeta-2, Zygin II, Zygin-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$354.00

$354.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$354.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$354.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FEZ2(9637)

General description

Fasciculation and elongation protein zeta-2 (FEZ2) is a homolog to the mammalian FEZ1, itself an ortholog of the C. elegans UNC-76. In contrast to FEZ1, FEZ2 mRNA is widely expressed in mouse tissues. FEZ2 interacts with protein kinase C (PKC)-zeta substrate and induces neurite extension of PC12 cells when co-expressed with a constitutively active form of PKC-zeta, suggesting FEZ2 may play an important role in the morphological changes of various cells by associating with PKC-zeta in a tissue non-specific manner. FEZ2 can interact with FEZ1 through its c-terminal regions and especially its coiled-coil region. At least two isoforms of FEZ2 are known to exist.

Immunogen

KLH-conjugated linear peptide corresponding to human FEZ2.

Application

Detect the FEZ2 protein using this Anti-FEZ2 validated for use in WB & IF.
Immunofluorescence Analysis: 5 µg/mLfrom a representative lot detected FEZ2 in human brain tissue.
Research Category
Neuroscience

Neuroscience
Research Sub Category
Developmental Neuroscience

Neuroregenerative Medicine

Quality

Evaluated by Western Blot in 3T3 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected FEZ2 in 15 µg of 3T3 cell lysate.

Target description

~38/50 kDa observed. An uncharacterized band may appear at ~48 kDa in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
3T3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service