Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN1446

Sigma-Aldrich

Anti-GPRIN1

from rabbit, purified by affinity chromatography

Synonym(s):

G protein-regulated inducer of neurite outgrowth 1, GRIN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GPRIN1(114787)

General description

G protein-regulated inducer of neurite outgrowth 1 (UniProt Q7Z2K8; also known as GRIN1) is encoded by the GPRIN1 (also known as KIAA1893) gene (Gene ID 114787) in human. Originally identified as a binding protein for guanosine 5′-3-O-(thio)triphosphate-bound G(z)alphaz, G protein-regulated inducer of neurite outgrowth 1 (GPRIN1) is a signaling molecule that functions as a downstream target of G(o)alpha. When co-expressed with constitutively active form of G(o)alpha, GPRIN1 is demonstrated to stimulate neurite extensions in Neuro2a cells. Gprin1 expression in mice is limited to the nervous system at all developmental stages, occurring within the entire embryonic mantle zones and becoming restricted to sets of nuclei at postnatal to adult stages. Gprin1 immunoreactivity is found colocalized with G(o)alpha at neuronal dendrites and axons, while being undetected in glial cells. Studies conducted in rat reveals that 7 nicotinic receptor ( 7) significantly reduces axon growth in hippocampal neurons by coupling to Gprin1, which targets it to the growth cone. RNAi-mediated Gprin1 knockdown abolishes the localization and calcium signaling of 7 at the growth cone. The 7/Gprin1 interaction appears intimately linked to a G(o)alpha, growth-associated protein 43, and CDC42 cytoskeletal regulatory pathway within the developing axon.

Specificity

This rabbit polyclonal antibody targets an internal sequence present in both spliced isoforms of human GPRIN1 protein reported by UniProt (Q7Z2K8).

Immunogen

KLH-conjugated linear peptide corresponding to an internal sequence of human GPRIN1.

Application

Anti-GPRIN1 Antibody, Cat. No. ABN1446, is a highly specific rabbit polyclonal antibody that targets GPRIN1 and has been tested in Immunohistochemistry and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected GPRIN1 in human prostate cancer, cerebral cortex and pancreas tissue sections.

Quality

Evaluated by Western Blotting in HT-29 cell lysate.

Western Blotting Analysis: A 1:20,000 dilution of this antibody detected GPRIN1 in 10 µg of HT-29 cell lysate.

Target description

~105 kDa observed. 102.4 kDa (isoform 1) and 80.95 kDa (isoform 2) calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service