Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE280

Sigma-Aldrich

Anti-Ars2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Serrate RNA effector molecule homolog, Arsenite-resistance protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRRT(51593)

General description

As a component of the nuclear RNA cap-binding complex (CBC), Ars2 is known to play a critical role in miRNA biogenesis by stabilizing and linking capped primary miRNA transcripts with the primary miRNA processing complex containing DROSHA and DGGR8. Unlike other CBC components, Ars2 expression also plays an important role in cell proliferation. In particular, depletion of Ars2 is shown to reduce the levels of several miRNAs, such as miR-21, let-7, and miR-155, which are implicated in cellular transformation.

Immunogen

Linear peptide corresponding to human Ars2.

Application

Immunoprecipitation Analysis: A representative lot from an independent laboratory detected Ars2 in IP. (Alex Achille, Department of Molecular Oncology, H. Lee Moffitt Cancer Center & Research Institute, Tampa, Florida, USA).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-Ars2 Antibody (Rabbit Polyclonal Antibody) validated in WB, IP to detect Ars2 also known as Serrate RNA effector molecule homolog, Arsenite-resistance protein 2.

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Ars2 in 10 µg of HEK293 cell lysate.

Target description

~120 kDa observed. Uncharacterized bands at ~78 kDa and 48 kDa may be observed in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service