AB752P
Anti-Collagen Type I Antibody
Chemicon®, from rabbit
About This Item
biological source
rabbit
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
chicken
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable
suitability
not suitable for Western blot
not suitable for immunohistochemistry (Paraffin)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... COL1A1(1277)
General description
Specificity
Application
Radioimmunoassay
ELISA
Not recommended for paraffin embedded tissue sections or Western blots. Optimal working dilutions must be determined by end user.
Cell Structure
ECM Proteins
Linkage
Physical form
Storage and Stability
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service