Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3802

Sigma-Aldrich

Anti-JAK1 Antibody

serum, Chemicon®

Synonym(s):

Janus Kinase 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$341.00

$341.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$341.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$341.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... JAK1(3716)

Specificity

Recognizes JAK1 (Janus Kinase 1). The members of the JAK family of cytoplasmic protein tyrosine kinases physically associate with ligand-bound receptors. It is this association that results in tyrosine phosphorylation and activation. JAK1 is approximately 130 kDa and contains a C-terminal tyrosine kinase domain, an adjacent kinase or kinase-related domain, and five other domains that are highly conserved among JAK family members.

Immunogen

Synthetic peptide corresponding to 21 amino acids of human/mouse/rat JAK1

(KTLIEKERFYESRCRPVTPS-C)

Application

Immunoprecipitation: Use at 5-10μl per 1-2mg lysate

Western blot: (1:1000-1:2000)

The amount of JAK1 in cell lines tends to be extremely low. If the signal is weak it is recommended that a RIPA cell lysate (1-2mg protein) be prepared and JAK1 be first concentrated by immunoprecipitation and then immunoblotted. A degradation product of JAK1 (~110kDa) is also detected by the antiserum in immunoblots.

Optimal working dilutions must be determined by end user.
Research Category
Signaling
Research Sub Category
Immunological Signaling
This Anti-JAK1 Antibody is validated for use in IP, WB for the detection of JAK1.

Physical form

Serum. Whole rabbit antiserum containing sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service