Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3075

Sigma-Aldrich

Anti-Aquaporin 7 Antibody

Chemicon®, from rabbit

Synonym(s):

AQP7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AQP7(364)

General description

Water is a critical component of all living cells. Interestingly, tissue membranes show a great degree of water permeability. Mammalian red cells, renal proximal tubules, and descending thin limb of Henle are extraordinarily permeable to water. Water crosses hydrophobic plasma membranes either by simple diffusion or through a facilitative transport mechanism mediated by special protein "aquaporin". Over the last decade, genes for several members of aquaporin family have been cloned, expressed, and their distribution studied in many tissues. Aquaporin-0 or MIP26 (major intrinsic protein 26 kDa), and Aquaporin-1 (purified from red cells) also called CHIP-28 (channel forming integral protein, 28 kDa; 268 AA; gene locus 7p14) has been the foundation of the growing family of aquaporins. The lens specific Aquaporin-0 represents up to 80% of total lens membrane protein. Defects in MIP26 are a cause of autosomal dominant cataract. The cataract Fraser mutation (CAT-FR or Shriveled) is a transposon-induced splicing error that substitutes a long terminal repeat sequence for the c-terminus of MIP. The lens opacity mutation (LOP) is an AA substitution that inhibits targeting of MIP to the cell membrane. Aquaporin-7 (rAQP7, 269aa) is abundantly expressed in testis. It is involved in water, glycerol and urea transport. Aquaporin-7 is also found in adipose tissue, kidney, and heart. Aquaporin-7 has been localized to in the spermatids and at maturing sperms. Aquaporin families of proteins are predicted to contain six transmembrane domains. The N and C-terminus are predicted to be cytoplasmic.

Specificity

Aquaporin 7. The immunogen peptide is unique to Aquaporin-7 without significant homology to any other Aquaporins.
The peptide has 81% similarity with the mouse Aquaporin-7

Immunogen

An 18 AA synthetic peptide within the N-terminal domain of rat Aquaporin-7 (Ishibashi et al. 1997) was selected for antibody production. This domain is predicted to be cytoplasmic.

Application

Detect Aquaporin 7 using this Anti-Aquaporin 7 Antibody validated for use in ELISA & WB.
Note: We recommend the use of 0.5-1.0% milk in all primary/secondary antibody incubations to suppress non-specific reactivity.

Western blot: 1-10 μg/mL using Chemiluminescence technique

ELISA: 0.5-1.0 μg/mL when tested against 1 μg/mL of the immunogen peptide.

Immunohistochemistry: We recommend using the affinity purified antibody at 2-10 μg/mL in paraformaldehyde fixed sections of tissues.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Physical form

Affinity Purified 1 mg/ml soln in PBS, pH 7.5 and 0.1% BSA and 0.05% sodium azide

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service