8.41853
2-Hydroxy-3-methoxybenzaldehyde
for synthesis
Synonym(s):
2-Hydroxy-3-methoxybenzaldehyde, o-Vanillin
About This Item
Recommended Products
Quality Level
assay
≥98.0% (GC)
mp
40-42 °C
transition temp
flash point 113 °C
InChI
1S/C8H8O3/c1-11-7-4-2-3-6(5-9)8(7)10/h2-5,10H,1H3
InChI key
JJVNINGBHGBWJH-UHFFFAOYSA-N
Application
Analysis Note
Melting range (lower value): ≥ 41 °C
Melting range (upper value): ≤ 45 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_c
113 °C - closed cup - (External MSDS)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service