Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41227

Sigma-Aldrich

N-(2-Acetamido)-iminodiacetic acid

for synthesis

Synonym(s):

N-(2-Acetamido)-iminodiacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2O5
CAS Number:
Molecular Weight:
190.15
MDL number:
UNSPSC Code:
12352106
EC Index Number:
247-530-0

Quality Level

form

powder

solubility

2.64 g/L

density

1.5257 g/cm3 at 20.2 °C

Storage temp.

2-30°C

InChI

1S/C6H10N2O5/c7-4(9)1-8(2-5(10)11)3-6(12)13/h1-3H2,(H2,7,9)(H,10,11)(H,12,13)/p-1

Inchi Key

QZTKDVCDBIDYMD-UHFFFAOYSA-M

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service