Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20920

Sigma-Aldrich

n-Octadecane

for synthesis

Synonym(s):

n-Octadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CH3
CAS Number:
Molecular Weight:
254.49
MDL number:
UNSPSC Code:
12352001
EC Index Number:
209-790-3

vapor pressure

<0.1 hPa ( 20 °C)

assay

≥99.0% (GC)

autoignition temp.

235 °C

bp

317 °C/1013 hPa

mp

26-29 °C

transition temp

flash point 154 °C

solubility

insoluble <0.0001 g/L

density

0.78 g/cm3 at 20 °C

InChI

1S/C18H38/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2/h3-18H2,1-2H3

InChI key

RZJRJXONCZWCBN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Identity (GC): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

330.8 °F - closed cup

flash_point_c

166 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service