8.20881
2-Nitroaniline
for synthesis
Synonym(s):
2-Nitroaniline, o-Nitraniline
About This Item
Recommended Products
Quality Level
assay
≥98.0% (GC)
form
powder
autoignition temp.
521 °C
potency
1600 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)
pH
6.1 (20 °C, 10 g/L in H2O, slurry)
mp
68-71 °C
transition temp
flash point 167 °C
bulk density
500 kg/m3
storage temp.
2-30°C
InChI
1S/C6H6N2O2/c7-5-3-1-2-4-6(5)8(9)10/h1-4H,7H2
InChI key
DPJCXCZTLWNFOH-UHFFFAOYSA-N
Analysis Note
Melting range (lower value): ≥ 70 °C
Melting range (upper value): ≤ 73 °C
Identity (IR): passes test
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2
target_organs
Blood
Storage Class
6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
332.6 °F - closed cup
flash_point_c
167.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service