Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.15027

Sigma-Aldrich

n-Decane-D22

deuteration degree min. 99% for NMR spectroscopy MagniSolv

Synonym(s):

n-Decane-D22

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D22
CAS Number:
Molecular Weight:
164.42
MDL number:
UNSPSC Code:
12142201

vapor pressure

1 hPa ( 16 °C)

form

liquid

autoignition temp.

205 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

0.7-5.4 % (v/v)

technique(s)

NMR: suitable

kinematic viscosity

<7 cSt(40 °C)

bp

174 °C

mp

-30 °C

transition temp

flash point 46 °C

density

0.845 g/cm3

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service