Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14681

Sigma-Aldrich

2,2,6,6-Tetramethylpiperidine-1-oxyl (free radical)

for synthesis

Synonym(s):

2,2,6,6-Tetramethylpiperidine-1-oxyl (free radical), Tempo

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18NO
CAS Number:
Molecular Weight:
156.25
MDL number:
UNSPSC Code:
12352119
EC Index Number:
219-888-8

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

assay

≥98.0% (GC)

form

powder

autoignition temp.

275 °C

reaction suitability

reagent type: oxidant

pH

8.3 (20 °C, 9 g/L in H2O)

mp

39-40 °C

transition temp

flash point 67 °C

solubility

9.7 g/L

density

0.912 g/cm3 at 40 °C

storage temp.

2-30°C

InChI

1S/C9H18NO/c1-8(2)6-5-7-9(3,4)10(8)11/h5-7H2,1-4H3

InChI key

QYTDEUPAUMOIOP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

pictograms

Corrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class

8A - Combustible, corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

152.6 °F - closed cup

flash_point_c

67 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service