8.07632
Rhodium(III) chloride hydrate (>38% Rh)
for synthesis
Synonym(s):
Rhodium(III) chloride hydrate (>38% Rh), Rhodium trichloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Cl3Rh · xH2O
CAS Number:
Molecular Weight:
209.26 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-165-4
Recommended Products
Quality Level
potency
753 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)
Analysis Note
Gehalt aus Cl (calc. on anhydrous substance): ≥ 96.0 %
Water (K. F.): 17.0 - 23.0 %
Water (K. F.): 17.0 - 23.0 %
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service