Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00952

1,2-Dibromoethane

for synthesis

Synonym(s):

1,2-Dibromoethane, Ethylene bromide, Ethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
187.86
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-444-5
Pricing and availability is not currently available.

vapor pressure

14.7 hPa ( 20 °C)

form

liquid

autoignition temp.

490 °C

potency

108 mg/kg LD50, oral (Rat)
300 mg/kg LD50, skin (Rabbit)

bp

132 °C/1013 hPa

mp

10 °C

solubility

4.04 g/L

density

2.18 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C2H4Br2/c3-1-2-4/h1-2H2

InChI key

PAAZPARNPHGIKF-UHFFFAOYSA-N

General description

for synthesis

Application

1,2-Dibromoethane for synthesis. CAS 106-93-4, chemical formula CH₂BrCH₂Br.

Analysis Note

Assay (GC, area%)≥ 99.0 % (a/a)Density (d 20 °C/ 4 °C)2.175 - 2.180Water (K. F.)≤ 0.10 %Identity (IR)passes testDue to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service