Skip to Content
MilliporeSigma
All Photos(1)

Documents

676485

Sigma-Aldrich

VEGFR2 Kinase Inhibitor II - CAS 288144-20-7 - Calbiochem

The VEGFR2 Kinase Inhibitor II, also referenced under CAS 288144-20-7, controls the biological activity of VEGFR2 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

VEGFR2 Kinase Inhibitor II - CAS 288144-20-7 - Calbiochem, ( Z)-5-Bromo-3-(4,5,6,7-tetrahydro-1 H-indol-2-ylmethylene)-1,3-dihydroindol-2-one, Vascular Endothelial Growth Factor Receptor 2 Kinase Inhibitor II, VEGFR Tyrosine Kinase Inhibitor XIII, (Z)-5-Bromo-3-(4,5,6,7-tetrahydro-1H-indol-2-ylmethylene)-1,3-dihydroindol-2-one, Vascular Endothelial Growth Factor Receptor 2 Kinase Inhibitor II, VEGFR Tyrosine Kinase Inhibitor XIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15BrN2O
CAS Number:
Molecular Weight:
343.22
UNSPSC Code:
12352200

assay

≥95% (HPLC)

form

solid

potency

70 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable and reversible indolin-2-one class of receptor tyrosine kinase (RTK) inhibitor [IC50 = 70 nM for VEGF-R2 (KDR/Flk-1), 920 nM for PDGF-Rβ, 4.92 µM for p60c-src, and 13.3 µM for FGF-R1]. The inhibition is suggested to be competitive with respective to ATP. Also inhibits VEGF-induced (IC50 = 110 nM) and PDGF-induced (IC50 = 2.01 µM) cell proliferation in umbilical vein endothelial cells (HUVEC). Does not exhibit any inhibitory effect on EGF-R kinase activity (IC50 >100 µM).
A membrane permeant and reversible indolin-2-one class of receptor tyrosine kinase (RTK) inhibitor [IC50 = 70 nM for VEGF-R2 (KDR/Flk-1), 920 nM for PDGF-Rβ, 4.92 µM for p60c-src, and 13.3 µM for FGF-R1]. The inhibition is suggested to be competitive with respect to ATP. Does not inhibit EGF-R kinase activity (IC50 >100 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
VEGF-R2 (KDR/Flk-1)
Product competes with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sun, L., et al. 2000. J. Med. Chem.43, 2655.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oisun Jung et al.
Journal of cell science, 132(20) (2019-09-29)
When targeted by the tumor-promoting enzyme heparanase, cleaved and shed syndecan-1 (Sdc1) then couples VEGFR2 (also known as KDR) to VLA-4, activating VEGFR2 and the directed migration of myeloma cells. But how VEGFR2 activates VLA-4-mediated motility has remained unknown. We now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service