The concentration of Lugol's Iodine for Microbiology, catalog number 624-71, is not explicitly stated. However, the SDS lists the concentration of both the I2 and KI as between 1 to 5%. It is likely that the product contains a "weak" iodine solution.
624
Lugol′s Iodine Solution
Harleco®, suitable for microbiology
Synonym(s):
Gram Stain
About This Item
Recommended Products
form
liquid
Quality Level
manufacturer/tradename
Harleco®
application(s)
diagnostic assay manufacturing
hematology
histology
microbiology
shipped in
ambient
storage temp.
15-25°C
General description
Application
For In-Vitro Diagnostic Use.
For Professional Use.
Lugol’s formulation of iodine which can be used for various Gram staining methods including Hucker and Stirling methods.
Legal Information
signalword
Warning
hcodes
pcodes
Hazard Classifications
STOT RE 2 Oral
target_organs
Thyroid
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the total iodine concentration of the 624-71 Lugol’s solution?
1 answer-
Helpful?
-
-
Are the compositions of 65042-93 and 1092611000 the same?
1 answer-
The composition of 624-71 and 1092611000 is considered proprietary, and while both are diluted solutions of iodine and potassium iodide, no statement can be made regarding their identical nature. Although both are used for the same purpose, there might be theoretical differences between the two products. It may be possible to substitute one for the other, but no side-by-side comparison studies have been performed to determine if one product can be substituted for the other.
Helpful?
-
-
Can Lugol's iodine solution 624-71 be used in staining parasite ova and protozoan cysts?
1 answer-
Lugol's Iodine solution (624-71) is formulated for use in staining bacteria in a modification of the Gram stain. Lugol's Iodine solutions available are intended for staining bacteria, not parasite ova and protozoan cysts.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service