Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.18216

Sigma-Aldrich

Oleic acid

purified grade

Synonym(s):

Oleic acid, cis-9-Octadecenoic acid, Elainic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
Beilstein/REAXYS Number:
1726542
MDL number:
UNSPSC Code:
12352211

grade

purified grade

vapor pressure

1 mmHg ( 176 °C)

refractive index

n20/D 1.459 (lit.)

bp

194-195 °C/1.2 mmHg (lit.)

mp

13-14 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-

InChI key

ZQPPMHVWECSIRJ-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service