Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

582005

Sigma-Aldrich

Anti-Telomerase (Ab-2) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-TERT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Product 582005 is not currently sold in your country. Contact Technical Service

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the ~125 kDa telomerase (TERT) protein.
Recognizes the ~125 kDa telomerase (TERT) protein.
This Anti-Telomerase (Ab-2) Rabbit pAb is validated for use in Frozen Sections, Immunoblotting, Immunprecipitation, Paraffin Sections for the detection of Telomerase (Ab-2).

Immunogen

Human
a synthetic peptide corresponding to amino acids from an internal region of the catalytic subunit of human telomerase reverse transcriptase (hTERT)

Application

Frozen Sections (1:250-1:1000)

Immunoblotting (1:100)

Immunprecipitation (1:40)

Paraffin Sections (1:250-1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Fu, W., et al. 2000. J. Mol. Neurosci.14, 3.
Kharbanda, S., et al. 2000. Curr. Biol. 10, 568.
Vaziri, H., et al. 1999. Mol. Cell. Biol. 19, 2373.
Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mauro Mattioli et al.
Cell biology international, 36(1), 7-19 (2011-09-02)
We set out to characterize stemness properties and osteogenic potential of sheep AEC (amniotic epithelial cells). AEC were isolated from 3-month-old fetuses and expanded in vitro for 12 passages. The morphology, surface markers, stemness markers and osteogenic differentiation were inspected
Daniel Reis Waisberg et al.
Clinics (Sao Paulo, Brazil), 67(9), 1039-1046 (2012-09-29)
This study sought to identify the relationship between fibroblast telomerase expression, myofibroblasts, and telomerase-mediated regulatory signals in idiopathic pulmonary fibrosis. Thirty-four surgical lung biopsies, which had been obtained from patients with idiopathic pulmonary fibrosis and histologically classified as usual interstitial
Florence Gizard et al.
Arteriosclerosis, thrombosis, and vascular biology, 31(2), 245-252 (2010-11-26)
Telomerase serves as a critical regulator of tissue renewal. Although telomerase activity is inducible in response to various environmental cues, it remains unknown whether telomerase is activated during the inflammatory remodeling underlying atherosclerosis formation. To address this question, we investigated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service