Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

567780

Millipore

SSC Buffer, 20X Powder Pack, ULTROL® Grade

Each pack provides an adequate amount of material to make 1 L of 20X concentrate that can be diluted to a 1X solution containing 150 mM NaCl, 15 mM sodium citrate, pH 8.0.

Synonym(s):

Saline-Sodium Citrate Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

sterile distilled water: 1%

shipped in

ambient

storage temp.

15-25°C

General description

Each pack provides an adequate amount of material to make 1 L of 20X concentrate that can be diluted to a 1X solution containing 150 mM NaCl, 15 mM sodium citrate, pH 8.0.
Each pack provides an adequate amount of material to prepare 1 liter of 20X buffer concentrate that can be diluted to a 1X solution, which will contain 150 mM NaCl and 15 mM sodium citrate, pH 7.85-8.15.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). 20X stock solutions are stable for up to 2 years at 4°C and 1X stock solutions are stable for up to 6 months at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service