Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553500

Sigma-Aldrich

RANTES, Human, Recombinant, E. coli

Synonym(s):

rhRANTES

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤100 pg/μg Endotoxin (pg/μg RANTES)

shipped in

ambient

storage temp.

−70°C

General description

Recombinant, human RANTES expressed in E. coli. RANTES (regulated on activation, normal T cell expressed and secreted) is a member of the β (C-C) chemokine family. Chemoattractant for unstimulated CD4+/CD45RO+ memory T cells. Induces Ca2+ mobilization in T cells. Acts as an antigen-independent activator of T cells in vitro.

Biochem/physiol Actions

ED₅₀ = 5-20 ng/ml as determined by the dose-dependent chemotaxis of THP-1 cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized, carrier free.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 3 months at -70°C.
Reconstitute with sterile dH₂O to a final concentration of 100-500 µg/ml. Further dilutions should be made in medium or buffered solution containing carrier protein, such as PBS with 0.1% BSA.

Other Notes

Bacon, K.B., et al. 1995. Science 269, 1727.
Devergne, O., et al. 1994. J. Exp. Med.179, 1689.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service