Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

552126

Millipore

Anti-Rab18 Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

General description

Recognizes the Rab18 protein.

Immunogen

recombinant rat brain Rab18

Application

ELISA (1:1000)

Immunoblotting (1:2000)

Warning

Toxicity: Standard Handling (A)

Other Notes

Does not cross-react with Cdc42, Rab1A, Rab3A, Rab5A, Rab6A, Rab8, Rab11B, Rab13, Rab15, Rab24, Rac1, or RhoB. Weakly with Rac1. Recognizes (weakly) uncharacterized proteins (~45 kDa and ~58 kDa) in HeLa cell extracts. Variables associated with assay conditions will dictate the proper working dilutions.
Lutcke, A., et al. 1994. J. Cell Sci.107, 3437.
Yu, H., et al. 1993. Gene132, 273.
Lang, V., et al. 1992. Plant Mol. Biol.21, 581.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service