Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539747

Sigma-Aldrich

InhibitorSelect 96-Well Tyrosine Kinase and Phosphatase Inhibitor Library IV - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 1 96-well plate(s)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−70°C

General description

The InhibitorSelect 96-Well Protein Tyrosine Kinase & Protein Tyrosine Phosphatase Inhibitor Library IV consists of 83 pharmacologically active, potent protein kinase and phosphatase inhibitors: most of the inhibitors included in this library are cell-permeable and reversible in their action. A majority of tyrosine kinase inhibitors included act in an ATP-competitive manner. The inhibitors in this library will be useful for target identification in drug discovery, biochemical pathway analysis, screening of new protein kinases/phosphatases, and other pharmaceutical-related applications. Supplied with a CD containing comprehensive documentation for each inhibitor. For more information please contact our technical service department.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

80 inhibitors are supplied in DMSO and 3 are supplied in H₂O, in the concentration range of 0.5-100 mM (50 µl/well).

Reconstitution

The contents of this pouch have been purged with nitrogen and are vacuum sealed. Cut open the pouch, take the plate out of the pouch, and allow it to thaw at room temperature. Please remove the lid gently. To re-store, purge with an inert gas, replace the lid tightly, place in a sealable bag, and store at -70°C in an upright position. Minimize freeze/thaw cycles. This product is stable for one year after receipt.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tracy Tran et al.
Cell stem cell, 29(7), 1083-1101 (2022-07-09)
Human pluripotent stem-cell-derived organoids are models for human development and disease. We report a modified human kidney organoid system that generates thousands of similar organoids, each consisting of 1-2 nephron-like structures. Single-cell transcriptomic profiling and immunofluorescence validation highlighted patterned nephron-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service