Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539672

Sigma-Aldrich

PKCδ, Human, Recombinant, S. frugiperda

Synonym(s):

Protein Kinase Cδ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥800 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Full-length, recombinant, human PKCδ expressed in S. frugiperda insect cells using a baculovirus expression system. PKC δ is classified as a novel PKC isozyme because it does not exhibit calcium dependence and it is activated in vitro by phorbol esters. Suitable for studies of PKCδ and its regulation. PKCδ is a Ca2+-independent and phospholipid-dependent enzyme.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will transfer 1 nmol of phosphate to PKCε Peptide Substrate (Cat. No. 539562) per min at 30°C, pH 7.5.

Physical form

In 250 mM NaCl, 20 mM HEPES, 5 mM DTT, 2 mM EDTA, 2 mM EGTA, 50% glycerol, 0.05% TRITON X-100 Detergent, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Hug, J. and Sarre, T.F. 1993. Biochem. J. 291, 329.
Kazanietz, M.G., et al. 1993. Mol.Pharmacol. 44, 298.
Koide, H., et al. 1992. Proc. Natl. Acad. Sci. USA 89, 1149.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service