539154
20S Proteasome, Rabbit
20S Proteasome is a catalytic core enzyme of the 26S proteasome that degrades ubiquitinated proteins. Fully functional with Z-LLVY-AMC as the substrate.
About This Item
Recommended Products
Quality Level
assay
≥95% (SDS-PAGE)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−70°C
General description
Biochem/physiol Actions
Packaging
Warning
Physical form
Reconstitution
Other Notes
Ciechanover, A. 1994. Cell 79, 13.
Dick, L.R., et al. 1994. J. Immunol. 152, 3884.
Dick, L.R., et al. 1991. Biochemistry 30, 2725.
Hough, R., et al. 1987. J. Biol. Chem. 262, 8303.
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service