Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.34342

Sigma-Aldrich

InhA Inhibitor, NITD-916

Synonym(s):

InhA Inhibitor, NITD-916, 6-((4,4-dimethylcyclohexyl)methyl)-4-hydroxy-3-phenylpyridin-2(1H)-one, Enoyl-acyl Carrier Protein Reductase Inhibitor, NITD-916, Enoyl-ACP reductase, NITD-916, 6-((4,4-dimethylcyclohexyl)methyl)-4-hydroxy-3-phenylpyridin-2(1H)-one , Enoyl-acyl Carrier Protein Reductase Inhibitor, NITD-916, Enoyl-ACP reductase, NITD-916

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25NO2
Molecular Weight:
311.42
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

General description

A cell-permeable, orally available, highly lipophilic 4-hydroxy-2-pyridone derivative that directly binds to the enoyl-substrate binding pocket of Mycobaterial enoyl reductase inhA in an NADH-dependent manner and inhibits its activity (IC50 = 570 nM). Shown to be about 5 to 8 times more potent ((MIC50 = 40 to 160 nM in multi-drug resistant strains of Mycobacterium) than isoniazid (MIC50 = 330 nM) and PA-824 (MIC50 = 400 nM). Exhibits in vivo efficacy in acute and established mouse models of Mycobacterium tuberculosis infection. Exhibits low to moderate metabolic clearance in mouse and human hepatic microsomes. Suggested to inhibit the fatty acid elongation step that blocks the biosynthesis of mycolic acids and weakens the cell wall mycolyl-arabinogalactan-peptidoglycan complex, and ultimately lysis of Mycobacterium.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Primary Target
InhA

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Manjunatha, U., et al. 2015. Sci. Trans. Med.7, 269.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service