Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

524733

Sigma-Aldrich

Phosphodiesterase 4D3, His•Tag®, Human, Recombinant, S. frugiperda

Synonym(s):

PDE4D3, cAMP-specific phosphodiesterase 4D isoform 3

Sign Into View Organizational & Contract Pricing

Select a Size

10 U
$497.00

$497.00


Estimated to ship onMay 10, 2025



Select a Size

Change View
10 U
$497.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

$497.00


Estimated to ship onMay 10, 2025


recombinant

expressed in insect cells (S. frugiperda)

Quality Level

form

liquid

specific activity

≥0.25 U/μL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Full-length, recombinant, human phosphodiesterase 4D3 (accession no. L20970) fused at the N-terminus to a His•Tag sequence and expressed in S. frugiperda insect cells using a baculovirus expression system. Supplied as a partially purified preparation.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 pmole cAMP to AMP per min at pH 7.5, 30°C.

Physical form

In 150 mM NaCl, 10 mM Tris, pH 8.3 .

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Gibson, L.C.D., et al. 2006. Eur. J. Pharmacol.538, 39.
Xu, R.X., et al. 2000. Science288, 1822.
Nemoz, G., et al. 1996. FEBS Lett.384, 97.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service