Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516640

Sigma-Aldrich

Phalloidin, Amanita phalloides - CAS 17466-45-4 - Calbiochem

Toxic bicyclic heptapeptide isolated from fungi.

Synonym(s):

Phalloidin, Amanita phalloides - CAS 17466-45-4 - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H48N8O11S
CAS Number:
Molecular Weight:
788.87
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 10 mg/mL
methanol: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C35H48N8O11S/c1-15-27(47)38-22-10-20-19-7-5-6-8-21(19)41-33(20)55-13-24(34(53)43-12-18(46)9-25(43)31(51)37-15)40-32(52)26(17(3)45)42-28(48)16(2)36-30(50)23(39-29(22)49)11-35(4,54)14-44/h5-8,15-18,22-26,41,44-46,54H,9-14H2,1-4H3,(H,36,50)(H,37,51)(H,38,47)(H,39,49)(H,40,52)(H,42,48)/t15-,16-,17-,18-,22-,23-,24-,25-,26+,35+/m0/s1

InChI key

KPKZJLCSROULON-QKGLWVMZSA-N

General description

Toxic bicyclic heptapeptide isolated from fungi. Useful as a potent and specific F-actin binding agent and inhibitor of F- to G-actin conversion.

Biochem/physiol Actions

Cell permeable: no
Primary Target
F-actin binding agent
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Mathews, J.B., et al. 1997. Am. J. Physiol. 272, C254.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service